Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ambp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_201118.2 | NP_957412.2 | 7 - 8 | 820 | 78 | |
GenBank mRNA | BC055598.1 | - | 7 - 8 | 693 | 78 | |
BC150464.1 | - | 7 - 8 | 679 | 78 | ||
BC165143.1 | - | 4 - 5 | 309 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons |