Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dnaja3b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_201342.1 | NP_958499.1 | 10 - 11 | 1329 | 133 | |
GenBank mRNA | BC055555.1 | - | 10 - 11 | 1329 | 133 |
Gene Symbol: LOC554962 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001123236.1 | NP_001116708.1 | 10 - 11 | 133 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |