Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dnaja3b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_201342.1 | NP_958499.1 | 7 - 8 | 979 | 73 | |
GenBank mRNA | BC055555.1 | - | 7 - 8 | 979 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |