Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dnaja3b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_201342.1 | NP_958499.1 | 8 - 9 | 1109 | 85 | |
GenBank mRNA | BC055555.1 | - | 8 - 9 | 1109 | 85 |
Gene Symbol: LOC554962 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001123236.1 | NP_001116708.1 | 8 - 9 | 1109 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |