Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: aldh9a1a.1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_201471.1 | NP_958879.1 | 7 - 8 | 972 | 76 | |
XM_005166774.3 | XP_005166831.1 | 7 - 8 | 1077 | 76 | ||
GenBank mRNA | BC045932.1 | - | 7 - 8 | 972 | 76 | |
BC066668.1 | - | 7 - 8 | 965 | 76 | ||
BC165753.1 | - | 6 - 7 | 935 | 76 | ||
BC166038.1 | - | 7 - 8 | 974 | 76 | ||
BC167472.1 | - | 7 - 8 | 974 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |