Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mmp13a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001290479.1 | NP_001277408.1 | 8 - 9 | 1259 | 125 | |
GenBank mRNA | BC049472.1 | - | 8 - 9 | 1180 | 125 | |
BC065591.1 | - | 8 - 9 | 1230 | 125 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |