Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mapk3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_201507.1 | NP_958915.1 | 3 - 4 | 742 | 85 | |
GenBank mRNA | AB030902.1 | - | 3 - 4 | 661 | 85 | |
AY922319.1 | - | 3 - 4 | 580 | 85 | ||
BC045505.1 | - | 3 - 4 | 742 | 85 | ||
BC066401.1 | - | 3 - 4 | 730 | 85 | ||
BC097073.1 | - | 3 - 4 | 815 | 85 | ||
BC163960.1 | - | 3 - 4 | 614 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Danio rerio Inventoried Probe spans exons |