Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ikzf5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_205601.1 | NP_991164.1 | 3 - 4 | 683 | 64 | |
XM_009293021.3 | XP_009291296.1 | 3 - 4 | 706 | 64 | ||
GenBank mRNA | AY394931.1 | - | 3 - 4 | 683 | 64 | |
BC078341.1 | - | 2 - 3 | 357 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |