Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: txn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_205641.1 | NP_991204.1 | 2 - 3 | 289 | 59 | |
GenBank mRNA | BC065316.1 | - | 2 - 3 | 289 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |