Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: barhl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_205740.1 | NP_991303.1 | 2 - 3 | 1182 | 69 | |
GenBank mRNA | AY145141.1 | - | 2 - 3 | 1182 | 69 | |
BC097030.1 | - | 2 - 3 | 1091 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |