Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: phox2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_207070.2 | NP_996953.2 | 1 - 2 | 324 | 87 | |
GenBank mRNA | BC066448.1 | - | 1 - 2 | 314 | 87 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |