Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: med8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_207083.1 | NP_996966.1 | 2 - 3 | 184 | 114 | |
GenBank mRNA | BC066472.1 | - | 2 - 3 | 184 | 114 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio Probe spans exons |