Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ttpal | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_212590.2 | NP_997755.2 | 4 - 5 | 791 | 151 | |
XM_021469732.1 | XP_021325407.1 | 4 - 5 | 826 | 151 | ||
GenBank mRNA | BC055264.1 | - | 4 - 5 | 780 | 151 | |
EH434754.1 | - | 4 - 5 | 800 | 151 | ||
EH460758.1 | - | 1 - 2 | 714 | 151 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Danio rerio Probe spans exons |