Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ufm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_212627.1 | NP_997792.1 | 3 - 4 | 154 | 140 | |
GenBank mRNA | AY394956.1 | - | 3 - 4 | 154 | 140 | |
BC044145.1 | - | 3 - 4 | 195 | 140 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio Probe spans exons |