Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: vapb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_212647.2 | NP_997812.1 | 2 - 3 | 470 | 77 | |
XM_021476783.1 | XP_021332458.1 | 2 - 3 | 419 | 77 | ||
GenBank mRNA | BC064648.1 | - | 398 | 77 | ||
BC152200.1 | - | 2 - 3 | 373 | 77 | ||
BC155104.1 | - | 2 - 3 | 381 | 77 | ||
EH552051.1 | - | 2 - 3 | 509 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons |