Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cul5a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | BC057475.1 | - | 14 - 15 | 1582 | 107 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio Probe spans exons |