Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cul5a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001328059.1 | NP_001314988.1 | 5 - 6 | 596 | 123 | |
GenBank mRNA | BC057475.1 | - | 5 - 6 | 567 | 123 | |
BC154320.1 | - | 5 - 6 | 560 | 123 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio Probe spans exons |