Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: grnb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_212738.1 | NP_997903.1 | 7 - 8 | 684 | 129 | |
XM_021470114.1 | XP_021325789.1 | 7 - 8 | 1007 | 129 | ||
GenBank mRNA | AY289606.1 | - | 7 - 8 | 684 | 129 | |
BC096854.1 | - | 7 - 8 | 678 | 129 | ||
BC163965.1 | - | 5 - 6 | 591 | 129 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |