Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: grnb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_212738.1 | NP_997903.1 | 9 - 10 | 908 | 105 | |
XM_021470114.1 | XP_021325789.1 | 9 - 10 | 1231 | 105 | ||
GenBank mRNA | AY289606.1 | - | 9 - 10 | 908 | 105 | |
BC096854.1 | - | 9 - 10 | 902 | 105 | ||
BC163965.1 | - | 7 - 8 | 815 | 105 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio Probe spans exons |