Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: yrk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001365553.1 | NP_001352482.1 | 6 - 7 | 683 | 97 | |
NM_212781.2 | NP_997946.1 | 6 - 7 | 683 | 97 | ||
GenBank mRNA | AY169369.1 | - | 6 - 7 | 643 | 97 | |
BC081616.1 | - | 6 - 7 | 683 | 97 | ||
FJ205701.1 | - | 6 - 7 | 630 | 97 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |