Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: smtnl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_212805.2 | NP_997970.1 | 8 - 9 | 1391 | 119 | |
GenBank mRNA | AB110459.1 | - | 8 - 9 | 1374 | 119 | |
BC090828.1 | - | 8 - 9 | 1395 | 119 | ||
BC165356.1 | - | 7 - 8 | 1242 | 119 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio Probe spans exons |