Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: yipf4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_212891.1 | NP_998056.1 | 4 - 5 | 590 | 112 | |
GenBank mRNA | BC066619.1 | - | 4 - 5 | 590 | 112 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |