Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: itm2bb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_212976.1 | NP_998141.1 | 5 - 6 | 752 | 88 | |
GenBank mRNA | BC053278.1 | - | 5 - 6 | 752 | 88 | |
BC155109.1 | - | 5 - 6 | 644 | 88 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Danio rerio Probe spans exons |