Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: blzf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001144803.1 | NP_001138275.1 | 5 - 6 | 927 | 89 | |
XM_021476915.1 | XP_021332590.1 | 6 - 7 | 964 | 89 | ||
GenBank mRNA | BC070020.1 | - | 5 - 6 | 871 | 89 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Danio rerio Probe spans exons |