Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sdf4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_213087.2 | NP_998252.1 | 4 - 5 | 929 | 99 | |
GenBank mRNA | BC049332.1 | - | 4 - 5 | 721 | 99 | |
BC066581.1 | - | 4 - 5 | 874 | 99 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |