Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cyb5a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_213135.2 | NP_998300.2 | 1 - 2 | 190 | 150 | |
GenBank mRNA | BC053263.1 | - | 1 - 2 | 177 | 150 | |
BC154824.1 | - | 1 - 2 | 190 | 150 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio Probe spans exons |