Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dnase1l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_213153.2 | NP_998318.2 | 6 - 7 | 626 | 108 | |
GenBank mRNA | BC056304.1 | - | 6 - 7 | 588 | 108 | |
CN022406.1 | - | 6 - 7 | 699 | 108 | ||
EH526021.1 | - | 3 - 4 | 539 | 108 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |