Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dnase1l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_213153.2 | NP_998318.2 | 8 - 9 | 883 | 117 | |
GenBank mRNA | BC056304.1 | - | 8 - 9 | 845 | 117 | |
EH526021.1 | - | 1 - 2 | 282 | 117 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |