Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sumo1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_213159.1 | NP_998324.1 | 4 - 5 | 335 | 68 | |
GenBank mRNA | BC056283.1 | - | 4 - 5 | 335 | 68 | |
BC067553.1 | - | 4 - 5 | 337 | 68 | ||
BC152217.1 | - | 4 - 5 | 271 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |