Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nudcd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_213191.1 | NP_998356.1 | 5 - 6 | 1013 | 81 | |
GenBank mRNA | BC065992.1 | - | 5 - 6 | 1013 | 81 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Danio rerio Probe spans exons |