Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gpm6aa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_213200.1 | NP_998365.1 | 5 - 6 | 722 | 94 | |
GenBank mRNA | AY070261.1 | - | 5 - 6 | 722 | 94 | |
BC064281.1 | - | 5 - 6 | 663 | 94 | ||
BC150455.1 | - | 5 - 6 | 746 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Danio rerio Probe spans exons |