Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dhps | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_213222.2 | NP_998387.1 | 2 - 3 | 337 | 80 | |
GenBank mRNA | BC047806.1 | - | 2 - 3 | 337 | 80 | |
BC066563.1 | - | 2 - 3 | 298 | 80 | ||
BC093128.1 | - | 2 - 3 | 337 | 80 | ||
BC164185.1 | - | 1 - 2 | 231 | 80 | ||
EH433453.1 | - | 2 - 3 | 366 | 80 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |