Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pnp5a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_213311.2 | NP_998476.1 | 2 - 3 | 416 | 59 | |
GenBank mRNA | BC066610.1 | - | 2 - 3 | 355 | 59 | |
EB931190.1 | - | 2 - 3 | 333 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |