Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tbx2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001102384.1 | NP_001095854.1 | 6 - 7 | 2025 | 83 | |
XM_005165556.4 | XP_005165613.1 | 6 - 7 | 2031 | 83 | ||
GenBank mRNA | BC068364.1 | - | 6 - 7 | 1997 | 83 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |