Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rbfox1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_214775.1 | NP_999940.1 | 3 - 4 | 504 | 96 | |
XM_005158173.4 | XP_005158230.1 | 3 - 4 | 556 | 96 | ||
XM_021466671.1 | XP_021322346.1 | 3 - 4 | 565 | 96 | ||
XM_021466672.1 | XP_021322347.1 | 4 - 5 | 538 | 96 | ||
XM_021466673.1 | XP_021322348.1 | 4 - 5 | 787 | 96 | ||
GenBank mRNA | AB074763.1 | - | 3 - 4 | 504 | 96 | |
BC163738.1 | - | 3 - 4 | 583 | 96 | ||
BC163747.1 | - | 3 - 4 | 583 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Danio rerio Probe spans exons |