Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rbfox1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_214775.1 | NP_999940.1 | 9 - 10 | 947 | 75 | |
XM_005158173.4 | XP_005158230.1 | 9 - 10 | 999 | 75 | ||
XM_021466671.1 | XP_021322346.1 | 9 - 10 | 1008 | 75 | ||
XM_021466672.1 | XP_021322347.1 | 10 - 11 | 981 | 75 | ||
XM_021466673.1 | XP_021322348.1 | 10 - 11 | 1230 | 75 | ||
GenBank mRNA | AB074763.1 | - | 9 - 10 | 947 | 75 | |
BC163738.1 | - | 9 - 10 | 1026 | 75 | ||
BC163747.1 | - | 9 - 10 | 1026 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons |