Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: asic1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_214790.2 | NP_999955.1 | 4 - 5 | 818 | 111 | |
XM_021478320.1 | XP_021333995.1 | 4 - 5 | 1076 | 111 | ||
GenBank mRNA | AJ609616.1 | - | 4 - 5 | 970 | 111 | |
BC162330.1 | - | 4 - 5 | 818 | 111 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |