Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rcan2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001320228.1 | NP_001307157.1 | 3 - 4 | 559 | 81 | |
NM_214819.2 | NP_999984.1 | 2 - 3 | 343 | 81 | ||
GenBank mRNA | BC070010.1 | - | 2 - 3 | 321 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |