Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rpl36a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001202510.1 | NP_001189439.1 | 3 - 4 | 345 | 82 | |
GenBank mRNA | AY099511.1 | - | 2 - 3 | 190 | 82 | |
BC055187.1 | - | 3 - 4 | 351 | 82 | ||
EH571835.1 | - | 2 - 3 | 636 | 82 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |