Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: fn3krp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | BC083230.1 | - | 6 | 1150 | 98 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Danio rerio Both primers and probe map within a single exon |