Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pcdh1a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024097.1 | NP_001019268.1 | 1 | 941 | 134 | |
GenBank mRNA | AY576977.1 | - | 1 | 941 | 134 | |
BC162531.1 | - | 1 | 939 | 134 | ||
BC162534.1 | - | 1 | 939 | 134 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Danio rerio Inventoried Both primers and probe map within a single exon |