Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: QKI | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081831.1 | NP_001075300.1 | 6 - 7 | 937 | 89 | |
XM_023632814.1 | XP_023488582.1 | 1118 | 89 | |||
XM_023632815.1 | XP_023488583.1 | 1097 | 89 | |||
XM_023632819.1 | XP_023488587.1 | 907 | 89 | |||
GenBank mRNA | AB177987.1 | - | 6 - 7 | 937 | 89 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Equus caballus Probe spans exons |