Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GAPDH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001163856.1 | NP_001157328.1 | 1 | 489 | 118 | |
GenBank mRNA | AF157626.1 | - | 3 - 4 | 310 | 118 | |
DQ403064.1 | - | 4 - 5 | 357 | 118 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Endogenous Controls Detect Genomic DNA Recommended Assay Equus caballus Inventoried Both primers and probe map within a single exon |