Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MYH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081759.1 | NP_001075228.1 | 21 - 22 | 2758 | 120 | |
XM_005597036.3 | XP_005597093.1 | 2862 | 120 | |||
GenBank mRNA | AB088366.1 | - | 21 - 22 | 2758 | 120 |
Set Membership: |
Amplicon length greater than or equal to 101 Equus caballus Probe spans exons |