Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MYH2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081760.1 | NP_001075229.1 | 20 - 21 | 2725 | 103 | |
XM_023651654.1 | XP_023507422.1 | 2839 | 103 | |||
XM_023651655.1 | XP_023507423.1 | 2821 | 103 | |||
GenBank mRNA | AB088365.1 | - | 20 - 21 | 2725 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 Equus caballus Probe spans exons |