Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ESR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081772.1 | NP_001075241.1 | 4 - 5 | 1302 | 64 | |
XM_023632844.1 | XP_023488612.1 | 1590 | 64 | |||
XM_023632845.1 | XP_023488613.1 | 1361 | 64 | |||
XM_023632846.1 | XP_023488614.1 | 1390 | 64 | |||
XM_023632847.1 | XP_023488615.1 | 1581 | 64 | |||
XM_023632848.1 | XP_023488616.1 | 1547 | 64 | |||
XM_023632849.1 | XP_023488617.1 | 1604 | 64 | |||
GenBank mRNA | AF124093.1 | - | 4 - 5 | 1302 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Equus caballus Probe spans exons |