Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PTGS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081775.2 | NP_001075244.1 | 2 - 3 | 291 | 85 | |
GenBank mRNA | AF027334.1 | - | 2 - 3 | 291 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Equus caballus Probe spans exons |