Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HTR2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081784.1 | NP_001075253.1 | 2 - 3 | 616 | 91 | |
XM_014732013.2 | XP_014587499.1 | 1530 | 91 | |||
XM_014732014.2 | XP_014587500.1 | 1443 | 91 | |||
GenBank mRNA | AB264327.1 | - | 2 - 3 | 616 | 91 | |
AY263355.1 | - | 2 - 3 | 233 | 91 |
Set Membership: |
Amplicon length between 86 and 100 G-protein coupled receptor 3' Most Equus caballus Probe spans exons |