Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OASL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081797.1 | NP_001075266.1 | 1 - 2 | 207 | 55 | |
GenBank mRNA | AY463162.1 | - | 1 - 2 | 207 | 55 |
Set Membership: |
Amplicon length less than or equal to 70 Equus caballus Probe spans exons |