Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TYRP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081840.2 | NP_001075309.2 | 1 - 2 | 463 | 106 | |
XM_014735365.2 | XP_014590851.1 | 615 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 Equus caballus Probe spans exons |